Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGGGTAGAGGGTGGGATTTAC[C/G]TCTTCCCAGAGCAGTGCCCTTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158371 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUC3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MUC3A - mucin 3A, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005960.1 | Intron | NP_005951.1 | ||||
XM_011516246.2 | Intron | XP_011514548.1 | ||||
XM_011516247.2 | Intron | XP_011514549.1 | ||||
XM_011516249.2 | Intron | XP_011514551.1 | ||||
XM_011516250.2 | Intron | XP_011514552.1 | ||||
XM_017012231.1 | Intron | XP_016867720.1 |
Set Membership: |
HapMap |