Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATTGTTTTTTTAGCAAAGTCCA[G/T]CCCTGCAGTCTAGCATTTGTAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616919 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FRMPD1 - FERM and PDZ domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014907.2 | Intron | NP_055722.2 | ||||
XM_011517803.2 | Intron | XP_011516105.1 | ||||
XM_011517804.2 | Intron | XP_011516106.1 | ||||
XM_011517805.2 | Intron | XP_011516107.1 | ||||
XM_011517806.2 | Intron | XP_011516108.1 | ||||
XM_017014480.1 | Intron | XP_016869969.1 | ||||
XM_017014481.1 | Intron | XP_016869970.1 | ||||
XM_017014482.1 | Intron | XP_016869971.1 |