Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCTAGATGGTCTCTAGGTGAGA[A/G]GAAATGAATTGGGTTAAAACATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603966 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR1C3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKR1C3 - aldo-keto reductase family 1, member C3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253908.1 | Intron | NP_001240837.1 | ||||
NM_001253909.1 | Intron | NP_001240838.1 | ||||
NM_003739.5 | Intron | NP_003730.4 |
Set Membership: |
HapMap |