Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAGACTCAAAGACATGCCAAGT[G/T]TGAGTAAAACCAGTCAGTCGGGAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rnf170 PubMed Links |
Rnf170 - ring finger protein 170 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029965.2 | Intron | NP_084241.1 |