Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCATTTCAAACATTCAAAATGC[A/G]GGTATTTTATACTAGTTTTTTTTTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nufip2 PubMed Links |
Nufip2 - nuclear fragile X mental retardation protein interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024205.2 | Intron | NP_001019376.1 |