Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTCCTCATTTTTCAGCCATTC[C/G]ACCTAAAGGCAGAAAAGGATGTGTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Unc5d PubMed Links |
Unc5d - unc-5 homolog D (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153135.3 | 865 | Silent Mutation | NP_694775.1 |