Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCCTAACACACGAAAGCACAAT[A/G]CACAAAGCTGCACACAGCATGAAGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cadm2 PubMed Links |
Cadm2 - cell adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145977.1 | Intron | NP_001139449.1 | ||||
NM_178721.4 | Intron | NP_848836.2 |