Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCCTGCCCCTGCAGCTGCCCTC[C/T]ATCTCTGTGCATCTGAGATAATGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rnaseh1 PubMed Links |
Rnaseh1 - ribonuclease H1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286865.1 | 1232 | UTR 3 | NP_001273794.1 | |||
NM_011275.3 | 1232 | UTR 3 | NP_035405.2 |