Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGGTGGGCTGGCAAGTGGAGG[A/G]AGGGCTAAGCAGAAGAGAGGGTAGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rbfox3 PubMed Links |
Rbfox3 - RNA binding protein, fox-1 homolog (C. elegans) 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024931.2 | Intron | NP_001020102.2 | ||||
NM_001039167.1 | Intron | NP_001034256.1 | ||||
NM_001039168.1 | Intron | NP_001034257.1 | ||||
NM_001285436.1 | Intron | NP_001272365.1 | ||||
NM_001285437.1 | Intron | NP_001272366.1 | ||||
NM_001285438.1 | Intron | NP_001272367.1 |