Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGGATTGTTGGGTTGTCTCTGT[A/G]GTTTGAGATTGTGAGAAGCAGAACC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Abca13 PubMed Links |
Abca13 - ATP-binding cassette, sub-family A (ABC1), member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178259.3 | Intron | NP_839990.2 |