Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAATAGAAACAGAGACTCTTTAT[G/T]AAGACACAGAAAGCACACCTGAGCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Zhx2 PubMed Links |
Zhx2 - zinc fingers and homeoboxes 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199449.2 | Intron | NP_955520.1 |