Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCGCTGATGGAAAGCCAACTAC[A/G]GTTCCCGCCAACCCCCACGGAGAAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Samd7 PubMed Links |
Samd7 - sterile alpha motif domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029489.3 | 867 | Silent Mutation | NP_083765.2 |