Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Btbd2 PubMed Links |
Btbd2 - BTB (POZ) domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145361.2 | Intron | NP_663336.2 |
Csnk1g2 - casein kinase 1, gamma 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |