Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCAATACAGAACAGGACTACA[C/T]GTGGTGACTGGGCACTGGGAACCTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Enpp2 PubMed Links |
Enpp2 - ectonucleotide pyrophosphatase/phosphodiesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136077.2 | 3183 | UTR 3 | NP_001129549.1 | |||
NM_001285994.1 | 3183 | UTR 3 | NP_001272923.1 | |||
NM_001285995.1 | 3183 | UTR 3 | NP_001272924.1 | |||
NM_015744.3 | 3183 | UTR 3 | NP_056559.2 |