Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTAAACGGTGGGTATCCCGGCCA[C/T]TGATTGAAGCTTGGAGTCCCAGGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Il3 PubMed Links |
Il3 - interleukin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010556.4 | 118 | Missense Mutation | NP_034686.2 |