Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGCGAAAGCTGCCTCTAACCCT[A/G]CAGGAAGTTATGTCTGGCCATCTAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gpc4 PubMed Links |
Gpc4 - glypican 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008150.2 | Intron | NP_032176.2 |