Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCACTCGGGGCTGGTTTCTCGGC[A/T]TGGGACTCTTTGCCTTTCGTGAGGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nrip1 PubMed Links |
Nrip1 - nuclear receptor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173440.2 | 2553 | Missense Mutation | NP_775616.1 |