Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTCAAACTCCTGCAAGGATTTT[C/T]TCTTCTGCAGCATGGGATATAATGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pacrg PubMed Links |
Pacrg - PARK2 co-regulated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027032.2 | Intron | NP_081308.1 |