Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGCAGCAGGAGGCTCTGGCCAG[C/G]GTAAGGGGTGAGGGGTGAGGCGGGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sec14l4 PubMed Links |
Sec14l4 - SEC14-like 4 (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_146013.1 | 118 | Missense Mutation | NP_666125.1 |