Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGTCCTTCTGGCTGCTCACAGG[A/G]ACCATCGGCTTCTACGCAGCCTACA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tm9sf4 PubMed Links |
Tm9sf4 - transmembrane 9 superfamily protein member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133847.3 | 2122 | Silent Mutation | NP_598608.2 |