Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ostn PubMed Links |
Ostn - osteocrin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Uts2b - urotensin 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198166.3 | Intron | NP_937809.1 |