Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTTTGAAAGTGTGGAAAGTACC[A/G]TGGAGAGTACGGTGGACAAATCAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fmnl2 PubMed Links |
Fmnl2 - formin-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172409.2 | 747 | Missense Mutation | NP_765997.2 |