Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGAACAAATTCCAAAAGCAGGT[C/T]TCTGAGTTCTCATACTTGACCCCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Col4a3 PubMed Links |
Col4a3 - collagen, type IV, alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007734.2 | Intron | NP_031760.2 |