Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCGCGCCTGTGGATTGCTGTAGC[C/T]GCTGCAATCGTGTTTGACAGGTTAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tiam1 PubMed Links |
Tiam1 - T cell lymphoma invasion and metastasis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145886.1 | Intron | NP_001139358.1 | ||||
NM_001145887.1 | Intron | NP_001139359.1 | ||||
NM_009384.3 | Intron | NP_033410.2 |