Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATAATTGTAAGAAGTATATAAAG[A/T]CTTGTGAAATTCAAGCCCAGTGCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Oxr1 PubMed Links |
Oxr1 - oxidation resistance 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130163.1 | Intron | NP_001123635.1 | ||||
NM_001130164.1 | Intron | NP_001123636.1 | ||||
NM_001130165.1 | Intron | NP_001123637.1 | ||||
NM_001130166.1 | Intron | NP_001123638.1 | ||||
NM_130885.2 | Intron | NP_570955.1 |