Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACAGAACCCTGCAGACTACCGTG[C/T]ACTTTAATCACATCTCTGTGACTGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ptprr PubMed Links |
Ptprr - protein tyrosine phosphatase, receptor type, R | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161837.1 | Intron | NP_001155309.1 | ||||
NM_001161838.1 | Intron | NP_001155310.1 | ||||
NM_001161839.1 | Intron | NP_001155311.1 | ||||
NM_001161840.1 | Intron | NP_001155312.1 | ||||
NM_011217.2 | Intron | NP_035347.1 |