Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fsbp PubMed Links |
Fsbp - fibrinogen silencer binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256142.1 | 345 | Silent Mutation | NP_001243071.1 |
Rad54b - RAD54 homolog B (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039556.3 | 345 | Intron | NP_001034645.1 | |||
NM_001256145.1 | 345 | Intron | NP_001243074.1 |