Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGAAGAGTCTCTAAACTTCAGC[A/G]TATGGCCTGCATTCTCTGGTAGCAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4933433G19Rik PubMed Links |
4933433G19Rik - RIKEN cDNA 4933433G19 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Ptdss1 - phosphatidylserine synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008959.3 | 2324 | UTR 3 | NP_032985.2 |