Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATAAAGGGGACTCCTCTTAAT[A/G]AGGGGAATCCCAGGATTGGACATCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pik3r5 PubMed Links |
Pik3r5 - phosphoinositide-3-kinase, regulatory subunit 5, p101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177320.2 | Intron | NP_796294.2 |