Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cmss1 PubMed Links |
Cmss1 - cms small ribosomal subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Tmem30c - transmembrane protein 30C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027651.1 | Intron | NP_081927.1 |