Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAACATAATCTAATTTCTGCTT[C/T]ATTGATGACTAAAAAGGTAGACATT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc27a6 PubMed Links |
Slc27a6 - solute carrier family 27 (fatty acid transporter), member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081072.1 | Intron | NP_001074541.1 |