Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGGGTAGAGTCATACTGGAACA[C/T]GTAGACCATGTAGTTGAGGTCAATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Srfbp1 PubMed Links |
Srfbp1 - serum response factor binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026040.3 | Intron | NP_080316.3 |