Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATACATAAGAAAAGAGAAATCTTT[C/T]TCCAAAGAAGACACAAAAAAGCCTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rbfox2 PubMed Links |
Rbfox2 - RNA binding protein, fox-1 homolog (C. elegans) 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110827.2 | Intron | NP_001104297.1 | ||||
NM_001110828.2 | Intron | NP_001104298.1 | ||||
NM_001110829.2 | Intron | NP_001104299.1 | ||||
NM_001110830.2 | Intron | NP_001104300.1 | ||||
NM_001286417.1 | Intron | NP_001273346.1 | ||||
NM_001286418.1 | Intron | NP_001273347.1 | ||||
NM_001286419.1 | Intron | NP_001273348.1 | ||||
NM_053104.6 | Intron | NP_444334.3 | ||||
NM_175387.3 | Intron | NP_780596.1 |