Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCTACAGAGATGATTTGTATTC[A/G]GTGGAGTAGGACCCAAAGTAGCGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Col25a1 PubMed Links |
Col25a1 - collagen, type XXV, alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244952.1 | Intron | NP_001231881.1 | ||||
NM_029838.4 | Intron | NP_084114.2 | ||||
NM_198711.3 | Intron | NP_942004.1 |