Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCCTAGAGACAGCTGCCCTGCA[C/T]GCCCTAGCCTTTAGCAATTGCCTGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc8a1 PubMed Links |
Slc8a1 - solute carrier family 8 (sodium/calcium exchanger), member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112798.2 | Intron | NP_001106269.1 | ||||
NM_001286684.1 | Intron | NP_001273613.1 | ||||
NM_011406.3 | Intron | NP_035536.2 |