Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATGGAGCACGGGTACCTTCAGC[A/G]TCCTGATGTGGCTCTCGGCTTCACT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cep128 PubMed Links |
Cep128 - centrosomal protein 128 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181815.3 | 2668 | Missense Mutation | NP_861536.3 |