Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCACTCCGAACATTGAGCTCATC[A/G]CTAGATATTATGTCAATGAGTTGGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Klhl20 PubMed Links |
Klhl20 - kelch-like 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039482.1 | 818 | Silent Mutation | NP_001034571.1 |