Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCCTCTGCAGGTTTGATTGAGA[C/T]GGCACATTTTGGATCTGTAAGCAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Zbtb10 PubMed Links |
Zbtb10 - zinc finger and BTB domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177660.3 | Intron | NP_808328.2 |