Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cd300lf PubMed Links |
Cd300lf - CD300 antigen like family member F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169153.1 | Intron | NP_001162624.1 | ||||
NM_145634.3 | Intron | NP_663609.2 |
Rab37 - RAB37, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163753.1 | Intron | NP_001157225.1 | ||||
NM_021411.4 | Intron | NP_067386.3 |