Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCACCTCTTAGAGCCACTGAGT[A/G]GTAGGAGGAGCTTACCTGGAAGGCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tns4 PubMed Links |
Tns4 - tensin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172564.3 | Intron | NP_766152.2 |