Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATGTGTAGGATATATATATAAA[A/G]TGAACACACTCCATGTAGTTAACAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sap130 PubMed Links |
Sap130 - Sin3A associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172965.2 | Intron | NP_766553.1 |