Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTCCCATGCTACAGCCATGGGGG[A/G]GGGGGGACGTAAATACCGTGAAAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Spag16 PubMed Links |
Spag16 - sperm associated antigen 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271533.1 | Intron | NP_001258462.1 | ||||
NM_025728.4 | Intron | NP_080004.1 | ||||
NM_029160.3 | Intron | NP_083436.2 |