Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGAAGGAATGTCCTGCCCTTT[A/C]CTTCTGTCTTTCTGTCTTCCCAGCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ddx31 PubMed Links |
Ddx31 - DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033294.3 | Intron | NP_001028466.2 |