Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAACTTCTGTTTGCCTGCCACTGT[C/T]TTGTGTGCTCTGCAGTGGTCTGTAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Arhgap18 PubMed Links |
Arhgap18 - Rho GTPase activating protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176837.2 | Intron | NP_789807.1 |