Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCAATAATCTAATTAAGGTTGA[C/T]GCATCACATCAAATTGAGGTAGAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Xirp2 PubMed Links |
Xirp2 - xin actin-binding repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024618.2 | 2444 | Silent Mutation | NP_001019789.1 | |||
NM_001083919.1 | 2444 | Silent Mutation | NP_001077388.1 |