Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pgk1 PubMed Links |
Pgk1 - phosphoglycerate kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Taf9b - TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001176.2 | Intron | NP_001001176.2 | ||||
NM_001167988.1 | Intron | NP_001161460.1 |