Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGTTCTCCTTTGAGGATTTCCTT[C/T]GGGGACCTCTCCATGTCTCCTCTTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rgs7bp PubMed Links |
Rgs7bp - regulator of G-protein signalling 7 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029879.2 | Intron | NP_084155.2 |