Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCGGAAATGGAGCAGATATCCC[G/C]GAGCTATTTGTTCCCATTGCTGGTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Peli2 PubMed Links |
Peli2 - pellino 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033602.2 | Intron | NP_291080.2 |