Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCAATACCAAATGATTTTTTCA[A/G]TATGGCTCCTGGCATTCAGTCCTCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Atp6v1h PubMed Links |
Atp6v1h - ATPase, H+ transporting, lysosomal V1 subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133826.4 | 879 | Missense Mutation | NP_598587.2 |