Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mir7664 PubMed Links |
Mir7664 - microRNA 7664 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Nsmf - NMDA receptor synaptonuclear signaling and neuronal migration factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039386.1 | Intron | NP_001034475.1 | ||||
NM_001039387.1 | Intron | NP_001034476.1 | ||||
NM_001177654.1 | Intron | NP_001171125.1 | ||||
NM_001177655.1 | Intron | NP_001171126.1 | ||||
NM_020276.3 | Intron | NP_064672.2 |
Pnpla7 - patatin-like phospholipase domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |